Learn More
S Murray and Co™ SAMCO™ Dissecting Scissors
Product Code. 12328099
Change view
Click to view available options
Length (Metric):
112 mm
125 mm
150 mm
Blade Style:
Curved
Straight
Unit Size:
Each
Pack of 2
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Blade Style Unit Size
12328099 112 mm Straight Each
31781820 112 mm Curved Pack of 2
12952055 125 mm Curved Each
12338099 125 mm Straight Each
12902055 150 mm Straight Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12328099 Supplier S Murray and Co™ Supplier No. E101/01
  • ONLINE EXCLUSIVE PRICE
    53.45 NOK
    22.37 NOK / Pack of 1
    Save 31.08 NOK
    58% Off
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
Use these scissors for a variety of applications. S Murray™ SAMCO™ Dissecting Scissors feature fine points, open shanks and stainless steel construction.
  • Stainless Steel dissecting scissors
Specifications
Product Type Dissecting Scissors
Blade Style Straight
Length (Metric) 112 mm
Material Stainless Steel