Learn More
Samarium, AAS standard solution, Specpure™ Sm 1000μg/mL
Product Code. 11366369
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 11366369
Brand: Thermo Scientific Alfa Aesar 088092.AP
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
MDL Number MFCD00011233
Specifications
Physical Form Liquid
Concentration Sm in 5% HNO3
UN Number UN3264
Quantity 500 mL
Grade Specpure
Chemical Name or Material Samarium, AAS standard solution, Sm 1000μg/mL