Learn More
Sam50 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28679-1-AP-150UL
Description
The Sam50 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Sam50. This antibody recognizes human, mouse, rat antigen. The Sam50 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| Sam50 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 25813 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |