Learn More
Salvequick Pilferproof Plaster Refill
Product Code. 12940475
Change view
Click to view available options
Material:
Fabric
Metal-detectable
Color:
Beige
Blue
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Color Unit Size
12940475 Metal-detectable Blue Each
12930475 Fabric Beige Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12940475 Supplier Safety First Supplier No. CD7
  • ONLINE EXCLUSIVE PRICE
    758.28 NOK
    518.12 NOK / Pack of 1
    Save 240.16 NOK
    31% Off
Estimated Shipment: 23-12-2026
Specifications
Material Metal-detectable
Product Type Salvequick Plaster Refill
Sterility Sterile
Quantity 6 Pk.
Packaging Type Refill
For Use With (Application) Salvequick Pilferproof Plaster Dispenser, Food Industry
Color Blue
Includes Twenty-one 72 x 19 mm; Fourteen 72 x 25 mm