missing translation for 'onlineSavingsMsg'
Learn More

Salmonella typhimurium Antibody (1E6cc) [Janelia Fluor™ 585], R&D Systems™

Product Code. 30171239
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30171239 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30171239 Supplier Novus Biologicals (Bio-Techne) Supplier No. NB11016952JF585
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Salmonella typhimurium Monoclonal antibody specifically detects Salmonella typhimurium in Bacteria samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Salmonella typhimurium
Applications ELISA
Classification Monoclonal
Clone 1E6cc
Conjugate Janelia Fluor 585
Formulation 50mM Sodium Borate
Gene Alias S. typhimurium;Salmonella LPS;Salmonellae
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Lipopolysaccharides (LPS) of S. typhimurium.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.