missing translation for 'onlineSavingsMsg'
Learn More

Salinomycin sodium salt, MedChemExpress

Product Code. 16403576
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
Product Code. Quantity unitSize
16403576 25 mg 25mg
16413367 50 mg 50mg
16407316 100 mg 100mg
3 options
This item is not returnable. View return policy

Product Code. 16403576

Brand: Medchem Express HY17439

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Salinomycin sodium salt (Salinomycin sodium), an antibiotic potassium ionophore, is a potent inhibitor of Wnt/β-catenin signaling. Salinomycin sodium salt (Salinomycin sodium) acts on the Wnt/Fzd/LRP complex, blocks Wnt-induced LRP6 phosphorylation, and causes degradation of the LRP6 protein. Salinomycin sodium salt (Salinomycin sodium) shows selective activity against human cancer stem cells.

Salinomycin sodium salt (Salinomycin sodium), an antibiotic potassium ionophore, is a potent inhibitor of Wnt/β-catenin signaling. Salinomycin sodium salt (Salinomycin sodium) acts on the Wnt/Fzd/LRP complex, blocks Wnt-induced LRP6 phosphorylation, and causes degradation of the LRP6 protein. Salinomycin sodium salt (Salinomycin sodium) shows selective activity against human cancer stem cells.

Specifications

Chemical Name or Material Salinomycin sodium salt
Physical Form Solid
CAS 55721-31-8
Color Light Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C42H69NaO11
For Use With (Application) Cancer-Kinase/protease
Synonym Salinomycin sodium Sodium salinomycin
Solubility Information DMSO : 100 mg/mL (129.37 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES [H][C@]1([C@](C)(CC2)O[C@]32[C@H](O)C=C[C@]4(O[C@]([H])([C@@H](CC)C([C@@H](C)[C@@H](O)[C@H](C)[C@]5([H])O[C@]([C@@H](CC)C(O[Na])=O)([H])CC[C@@H]5C)=O)[C@@H](C)C[C@H]4C)O3)CC[C@@](CC)(O)[C@H](C)O1
Molecular Weight (g/mol) 772.98
Formula Weight 772.98
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.