Learn More
Scharlab SALICYLSYRA PSS 500G
Brand:  Scharlab AC20020500
SALICYLSYRA PSS 500G
Product Code. 11382455
  • 1176.00 NOK / 500g
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Salicylic acid, extra pure, Pharmpur, Ph Eur, BP, USP
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .