Learn More
Salicyloyl Phytosphingosine, TRC
Specifications
Specifications
| Chemical Name or Material | Salicyloyl Phytosphingosine |
| CAS | 212908-67-3 |
| Molecular Formula | C25 H43 N O5 |
| Quantity | 2.5 mg |
| Synonym | Salicyloyl Phytosphingosine, N-[(1S,2S,3R)-2,3-Dihydroxy-1-(hydroxymethyl)heptadecyl]-2-hydroxybenzamide |
| InChI Formula | InChI=1S/C25H43NO5/c1-2-3-4-5-6-7-8-9-10-11-12-13-18-23(29)24(30)21(19-27)26-25(31)20-16-14-15-17-22(20)28/h14-17,21,23-24,27-30H,2-13,18-19H2,1H3,(H,26,31)/t21-,23+,24-/m0/s1 |
| SMILES | CCCCCCCCCCCCCC[C@@H](O)[C@@H](O)[C@H](CO)NC(=O)c1ccccc1O |
| IUPAC Name | 2-hydroxy-N-[(2S,3S,4R)-1,3,4-trihydroxyoctadecan-2-yl]benzamide |
| Molecular Weight (g/mol) | 437.613 |
| Formula Weight | 437.314 |