missing translation for 'onlineSavingsMsg'
Learn More

Salicylidene salicylhydrazide, 97%, Thermo Scientific™

Product Code. p-7044789
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11358055 5 g 5g
11368055 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11358055

Brand: Thermo Scientific Alfa Aesar L08235.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3232-36-8
Molecular Formula C14H12N2O3
Molecular Weight (g/mol) 256.26
MDL Number MFCD00043494
InChI Key NCCJLHWEUIAECU-MDZDMXLPSA-N
Synonym salicylidene salicylhydrazide, benzoic acid, 2-hydroxy-, 2-hydroxyphenyl methylene hydrazide, benzoic acid, 2-hydroxy-, 2-2-hydroxyphenyl methylene hydrazide, 2-hydroxy-n'-1e-6-oxocyclohexa-2,4-dien-1-ylidene methyl benzohydrazide, scs, 2-hydroxybenzylidene salicylohydrazide, tocris-2143, 2-hydroxy-n'-e-2-hydroxyphenyl methylidene benzohydrazide, n-1e-2-2-hydroxyphenyl-1-azavinyl 2-hydroxyphenyl carboxamide
PubChem CID 5483116
IUPAC Name 2-hydroxy-N'-[(E)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]benzohydrazide
SMILES OC1=CC=CC=C1C(=O)NN\C=C1/C=CC=CC1=O

Specifications

Melting Point 280°C to 284°C
Quantity 5 g
Beilstein 2944701
Formula Weight 256.26
Percent Purity 97%
Chemical Name or Material Salicylidene salicylhydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.