Learn More
Salicylic Acid, Sigma-Aldrich™
Product Code. 15836600
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15836600 1 kg 1kg
15826600 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15836600 Supplier Sigma-Aldrich Supplier No. 8187311000
 Restricted Item
Estimated Shipment: 28-01-2027
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69-72-7
Specifications
Melting Point 157°C to 159°C
Boiling Point 211°C/1013 hPa
Density 1.443 g/cm3 at 20°C
Quantity 1 kg
MDL Number MFCD00002439
Synonym Salicylic acid, 2-Hydroxybenzoic acid, Salicylic acid
Molecular Weight (g/mol) 138.12
Grade Ungraded
Chemical Name or Material Salicylic Acid