missing translation for 'onlineSavingsMsg'
Learn More

Salicylamide, MedChemExpress

Product Code. p-200099118
Encompass_Preferred
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 16418638

Brand: Medchem Express HYB0811

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Salicylamide is an inhibitor of microsomal UDP-glucuronosyltransferase. Salicylamide is an analgesic and anti-pyretic agent.

Salicylamide is an inhibitor of microsomal UDP-glucuronosyltransferase. Salicylamide is an analgesic and anti-pyretic agent.

Specifications

Chemical Name or Material Salicylamide
CAS 65-45-2
Color White
Purity Grade Notes Research
Molecular Formula C7H7NO2
Quantity 10 g
Synonym 2-Hydroxybenzamide
Solubility Information DMSO : ≥ 100 mg/mL (729.18 mM) ∣H2O : 0.1 mg/mL (0.73 mM; Need ultrasonic)
SMILES O=C(N)C1=CC=CC=C1O
Molecular Weight (g/mol) 137.14
Formula Weight 137.14
Percent Purity 98.88%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.