Learn More
Salicylaldehyde, TRC
Product Code. 30133812
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133812 100 g 100g
30138342 500 g 500g
30040448 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133812 Supplier Toronto Research Chemicals Supplier No. TRCS088145100G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 90-02-8
Molecular Formula C7 H6 O2
Molecular Weight (g/mol) 122.12
Synonym Salicylaldehyde, Benzaldehyde, 2-hydroxy-, Salicylaldehyde (8CI), 2-Hydroxybenzaldehyde, 2-Formylphenol, 2-Hydroxy-1-benzaldehyde, NSC 112278, NSC 49178, NSC 83559, NSC 83560, NSC 83561, NSC 83562, NSC 97202, Salicylal, Salicylic aldehyde, o-Formylphenol, o-Hydroxybenzaldehyde
IUPAC Name 2-hydroxybenzaldehyde
SMILES Oc1ccccc1C=O
Spezifikation
Quantity 100 g
InChI Formula InChI=1S/C7H6O2/c8-5-6-3-1-2-4-7(6)9/h1-5,9H
Formula Weight 122.0368
Percent Purity >95%
Chemical Name or Material Salicylaldehyde
Grade HPLC