Learn More
Saint-Gobain Versilon™ Plasticizer-Free Tubing, Formulation 2001
Product Code. 11740418
Change view
Click to view available options
Diameter (English) Inner:
1/16 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/8 in.
Diameter (English) Outer:
1 in.
1/2 in.
1/4 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
7/16 in.
7/8 in.
Thickness (English):
1/16 in.
1/8 in.
Unit Size:
15m
Product Code. Diameter (English) Inner Diameter (English) Outer Thickness (English) unitSize
11740418 3/4 in. 1 in. 1/8 in. 15m
11780408 3/16 in. 5/16 in. 1/16 in. 15m
11700418 5/16 in. 7/16 in. 1/16 in. 15m
11790408 1/4 in. 3/8 in. 1/16 in. 15m
11760408 1/16 in. 3/16 in. 1/16 in. 15m
11770408 1/8 in. 1/4 in. 1/16 in. 15m
11710418 3/8 in. 1/2 in. 1/16 in. 15m
11720418 1/2 in. 3/4 in. 1/8 in. 15m
11730418 5/8 in. 7/8 in. 1/8 in. 15m
9 options
Product Code. 11740418
Brand: Saint-Gobain 9643016
Only null left
Add to basket
Add to basket
Ideal for use in peristaltic pump systems, soap and detergent dispensing, ink transfer, water purification lines, chemical transfer, and food and beverage applications
Versilon™ Plasticizer-Free Tubing, Formulation 2001 is uniquely engineered to provide flexibility, chemical resistance and extended pump life in a clear, plasticizer-/oil-free tubing product.
  • Does not contaminate fluids
  • Superior flex life in peristaltic pumps
  • Chemically resistant to a wide range of fluids
  • Crystal clear for easy visual flow monitoring
  • Meets FDA criteria for food contact
  • Will not degrade over time
  • Will not release hazardous byproducts when properly incinerated
  • Does not yield any taste
  • Helps ensure accurate results from analytical instrument tests
Order Info
All products sold in 50-foot (15.24m) lengths
Compliance
Compliant with FDA 21 CFR, 177.2600
†At 73°F (23°C).
Specifications
Certifications/Compliance Compliant with FDA 21 CFR, 177.2600
Diameter (English) Inner 3/4 in.
Diameter (Metric) Inner 19.1 mm
Diameter (English) Outer 1 in.
Diameter (Metric) Outer 25.4 mm
Overall Length (English) 50 ft.
Thickness (English) 1/8 in.
Thickness (Metric) 3.2 mm