missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain Tygon™ High-Purity Pressure Tubing

Product Code. 11724688
Click to view available options
Overall Length (English):
50 ft.
Unit Size:
15m
This item is not returnable. View return policy

Product Code. 11724688

Brand: Saint-Gobain 9536050

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity pressure tubing with low sorption, flexibility and clarity, available in a single tubing

  • Extremely low sorption to aqueous fluids
  • Plasticizer free
  • Tough braid reinforcement for elevated working pressures
  • Meets USP Class VI and FDA Criteria

Specifications

Autoclavable Autoclavable
Material Tygon
Overall Length (English) 50 ft.
Maximum Pressure 150 psi
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.