Learn More
Saint-Gobain Tygon™ Formula 2475 High-Purity Tubing
Product Code. 12356919
Change view
Click to view available options
Diameter (English) Inner:
0.06 in.
0.18 in.
1/2 in.
1/4 in.
3/8 in.
5/16 in.
Diameter (English) Outer:
0.18 in.
0.31 in.
0.37 in.
0.43 in.
1/2 in.
3/4 in.
Thickness (English):
0.06 in.
0.12 in.
Unit Size:
15m
Each
Product Code. Diameter (English) Inner Diameter (English) Outer Thickness (English) unitSize
12356919 1/4 in. 0.37 in. 0.06 in. Each
11724678 0.06 in. 0.18 in. 0.06 in. 15m
12326919 0.06 in. 0.18 in. 0.06 in. Each
11734678 0.18 in. 0.31 in. 0.06 in. 15m
12366919 5/16 in. 0.43 in. 0.06 in. Each
11754678 5/16 in. 0.43 in. 0.06 in. 15m
11774678 1/2 in. 3/4 in. 0.12 in. 15m
12376919 3/8 in. 1/2 in. 0.06 in. Each
11744678 1/4 in. 0.37 in. 0.06 in. 15m
9 options
Product Code. 12356919
Brand: Saint-Gobain ACG00017
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Environmentally friendly—safe to dispose of through incineration
  • Contains no plasticizers that can leach into or contaminate fluid, yet tubing remains flexible
  • Virtually no absorption of key fluid constituents into tubing material or fluid adsorption onto tubing walls; fluid alteration and loss is limited
  • Ideal for handling sensitive fluids such as pharmaceutical or biological solutions
  • Also available braid-reinforced to withstand solutions with elevated working pressure (Contact your Fisher Scientific Customer Service Representative for details)
Order Info
In packs of 50 ft. (15m)
Compliance
Meets USP Class VI, FDA and NSF specifications. Documented biocompatibility to ISO 10993 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets USP Class VI, FDA and NSF specifications. Documented biocompatibility to ISO 10993 standards.
Diameter (English) Inner 1/4 in.
Diameter (Metric) Inner 6.4 mm
Material Tygon
Diameter (English) Outer 0.37 in.
Diameter (Metric) Outer 9.5 mm
Overall Length (English) 50 ft.
Overall Length (Metric) 15 m
Thickness (English) 0.06 in.
Thickness (Metric) 1.6 mm