Learn More
Saint Gobain Life Sciences Chemware™ Raschig Rings
Product Code. 12301409
Change view
Click to view available options
Diameter (Metric) Outside:
3 mm
6 mm
Length (Metric):
3 mm
6 mm
Unit Size:
Pack of 300
Pack of 500
Product Code. Diameter (Metric) Outside Length (Metric) unitSize
12301409 6 mm 6 mm Pack of 300
10575434 3 mm 3 mm Pack of 500
2 options
Product Code. 12301409
Brand: Saint Gobain Life Sciences D1069134
Chemware™ Raschig Rings for use with distillation pumps in chemical processes and liquid or gas transfer applications.
  • Packing for distillation columns and scrubbers.
  • Chemically inert except for molten alkali metals, fluorine, and strong fluorinating agents.
  • High surface area per unit of volume.
  • Minimum resistance to gas and liquid flow.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For chemical processes and liquid or gas transfer applications
Length (Metric) 6 mm
Type Raschig Ring
Material PTFE
For Use With (Equipment) For distillation columns
Diameter (Metric) Outside 6 mm
Length (English) 0.2 in.
Diameter (English) Outside 0.2 in.