Learn More
Saint-Gobain Chemware™ PTFE Fluoropolymer Resin Policemen
Description
- Will not scratch or damage delicate surfaces
- Choose from Hayman Style, Double Flat End Style, Double-ended Lab Spoon Style and Tapered Pallet with reinforced Hardwood Handle
- Extra heavy-duty PTFE coatings
- Resistant to low and high temperatures
- Use for handling chemically active materials
- Corrosion resistant
- Non-wettable
- Chemically inert
Specifications
Specifications
| Product Type | Spatula, Policemen |
| For Use With (Application) | Delicate transfer or scraping of precipitates and residues |
| Length (English) | 4 in. |
| Diameter (English) Outer | 0.28 in. |
| Diameter (Metric) Outer | 0.023 cm |
| Includes | Chisel tip and diagonal cut end |
| Material | PTFE Fluoropolymer Resin |
| Length (Metric) | 10 cm |