Learn More
MEDICOM Safewear Low Waterproof Overboots with Sole
Product Code. 31781294 Shop All Medicom Products
Change view
Click to view available options
Size:
Universal
Unit Size:
Pack of 400
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781294 Universal Pack of 400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31781294 Supplier MEDICOM Supplier No. 10.202
  • ONLINE EXCLUSIVE PRICE
    2899.48 NOK
    2410.20 NOK / Pack of 400
    Save 489.28 NOK
    16% Off
Estimated Shipment: 01-12-2026
Low waterproof overshoes provide total protection against moisture and grip, even on wet floors.
  • Made of embossed PE, the overboot is 100% waterproof: tested according to the EN ISO 811 water penetration resistance method under pressure
  • The sole provides good grip on the ground, even on wet surfaces
  • Excellent abrasion resistance even during prolonged use: tested according to the EN ISO 12947-2 method
  • Reinforced model in 200 μm embossed PE
  • Its foot-shaped (anatomical) design ensures easy fitting for the user, even in uncomfortable positions
  • The overboot fully covers the user's footwear, fitting sizes from 34 to 48
  • The elastic band ensures excellent ankle support
Specifications
Color White
Material Polyethylene
Size Universal
Gender Unisex
Waterproof Yes
Product Type Overboot