Learn More
HALYARD™ SAFEVIEW™ Protective Eyewear, Replacement Lenses
Shop All Halyard Products
Click to view available options
:
Replacement lenses
Description
- HALYARD™ SAFEVIEW™ Eyewear provides reliable eye protection for healthcare professionals and other users in environments where splashing or spraying may occur.
- For use in situations which may involve splashing or spraying
- Certified as PPE Category II, for moderate risks
- Lightweight design to reduce strain and increase comfort in use
- Colorful frames appeal to a variety of preferences
- Designed to be easily worn over most prescription glasses
- Constructed from natural rubber latex-free and DEHP-free materials
- Lenses film: clear polyester; Frames: polypropylene
Specifications
Specifications
| Certifications/Compliance | PPE Regulation 2016/425 (PPE category II) |
| Color | Clear |
| Product Type | Eyeshield Lens |
| No. per Case | 250 |
| Quantity | 250 |
| No. per Pack | 25 |
| Size | One-size |
| Disposable | Single-use |
| Autoclavable | Not Autoclavable |