Learn More
Sartorius™ Safety Weighing Cabinets
Product Code. 15344494 Shop All Sartorius Products
Change view
Click to view available options
Flow Rate:
.057 m3/sec. w/ direction connection, .062 m3/sec. w/ connection via fan filter
.066 m3/sec. w/ direction connection, .072 m3/sec. w/ connection via fan filter
.076 m3/sec. w/ direction connection, .083 m3/sec. w/ connection via fan filter
.089 m3/sec. w/ direction connection, .095 m3/sec. w/ connection via fan filter
Width (Metric):
890 mm
1230 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Width (Metric) Unit Size
15344494 .066 m3/sec. w/ direction connection, .072 m3/sec. w/ connection via fan filter 890 mm Each
10309444 .089 m3/sec. w/ direction connection, .095 m3/sec. w/ connection via fan filter 1230 mm Each
10762817 .076 m3/sec. w/ direction connection, .083 m3/sec. w/ connection via fan filter 1230 mm Each
15333461 .057 m3/sec. w/ direction connection, .062 m3/sec. w/ connection via fan filter 890 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15344494 Supplier Sartorius™ Supplier No. SWC900T
  • ONLINE EXCLUSIVE PRICE
    309360.00 NOK
    280847.85 NOK / Pack of 1
    Save 28512.15 NOK
    9% Off
 Restricted Item
Estimated Shipment: 14-01-2027
Ensure a safe working environment for laboratory staff without sacrificing ergonomics or precision in weighing. Sartorius™ Safety Weighing Cabinets offer important advantages in design and construction over traditional laboratory hoods.
  • The balance and weighing cabinet are perfectly matched to each other
  • Maximum personnel protection: contained area around the laboratory balance which prevents any air or finely powdered particles from escaping into the operating personnel´s work area
  • Reliable weighing results: air current practically turbulence-free, and, consistent and repeatable weighing results.
  • Clear acrylic glass construction with perfect view of the balance and the sample to be weighed
  • Extremely stable base made of black granite absorbs vibrations
  • Contaminated materials can be disposed of safely in a double-bag system
  • Calibrated airflow alarm system with audit trail function sounds an alarm whenever the airflow velocity drops below a defined level
TRUSTED_SUSTAINABILITY
Specifications
Material Acrylic Glass
Air Supply Constant
Depth (Metric) 750 mm
Flow Rate .066 m3/sec. w/ direction connection, .072 m3/sec. w/ connection via fan filter
For Use With (Application) Ensure a Safe Working Environment for Laboratory Staff Without Sacrificing Ergonomics or Precision in Weighing
Height (Metric) 770 mm
Width (Metric) 890 mm