Learn More
Sartorius Safety Weighing Cabinet without Filtration
Product Code. 16363086 Shop All Sartorius Products
Change view
Click to view available options
Width (Metric):
500 mm
750 mm
Length (Metric):
890 mm
900 mm
Unit Size:
Each
Product Code. Width (Metric) Length (Metric) unitSize
16363086 750 mm 900 mm Each
16687293 500 mm 890 mm Each
2 options
Product Code. 16363086
Brand: Sartorius SWC900TNF
This item requires a price update prior to being ordered.
Request a Price for this item here.
Safety Weighing Cabinets ensure a safe working environment for laboratory staff without sacrificing ergonomics or precision in weighing. Personnel safety and reliable weighing results from a single source.
  • Appropriate protective measures must be taken to protect the health of all laboratory staff when they weigh hazardous compounds in powdered form.
  • The Sartorius SWC Safety Weighing Cabinet offers important advantages in design and construction over traditional laboratory hoods:
    • The SWC creates a contained area around the laboratory balance which prevents any air or finely powdered particulates from escaping into the operating personnel's work area.
    • At the same time, the constant inlet air velocity and the special cabinet construction both keep the air current practically turbulence-free, and, as a result, ensure consistent and repeatable weighing results.
    • The balance and weighing cabinet are perfectly matched to each other.
    • With its SWC Safety Weighing Cabinet, Sartorius has succeeded in uniting two otherwise contradictory requirements: maximum personnel protection and reliable weighing results.
    • Maximum personnel protection: Contained area around the laboratory balance which prevents any air or finely powdered particles from escaping into the operating personnel's work area
    • Reliable weighing results: Air current practically turbulence-free, and, consistent and repeatable weighing results.
    • Clear acrylic glass construction with perfect view of the balance and the sample to be weighed
    • Extremely stable base made of black granite absorbs vibrations
    • Contaminated materials can be disposed of safely in a double-bag system
    • Calibrated airflow alarm system with audit trail function sounds an alarm whenever the airflow velocity drops below a defined level
TRUSTED_SUSTAINABILITY
Specifications
Material Acrylic Glass
Length (Metric) 900 mm
Width (English) 29.5 in.
Width (Metric) 750 mm