Learn More
Sartorius™ Safety Weighing Cabinet
Artikelnummer. 15680835 Alle ansehen Sartorius Produkte
Change view
Click to view available options
Material:
Acrylic Glass
Packungsgröße:
Each
Artikelnummer. Material unitSize
15680835 Acrylic Glass Each
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15680835
Marke: Sartorius™ SWC1200TNF
This item requires a price update prior to being ordered.
Request a Price for this item here.
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Ensures a safe working environment for lab staff without sacrificing ergonomics or precision in weighing.
  • Personnel safety and reliable weighing results from a single source
  • Without filter
  • 1230 x 750 x 770 mm
  • Clear acrylic glass construction affords a perfect view of the balance and sample
  • Excellent for working with extremely small samples
  • All surfaces easy to clean and compatible with conventional cleaning agents
  • Extremely stable base of black granite absorbs vibrations that can affect the balance
  • Creates a contained area around the balance which prevents any air or finely powdered particulates from escaping into the work area
  • Includes data-logging alarm, lighting unit, waste disposal system, airflow smoke test kit and antistatic cleaning wipes
TRUSTED_SUSTAINABILITY
Spezifikation
Product Type Safety Weighing Cabinet
Material Acrylic Glass
Depth (Metric) 750 mm
For Use With (Application) Safe weighing
Height (Metric) 770 mm
Width (Metric) 1230 mm