Learn More
3M™ Safety-Walk™ Slip-Resistant Fine Resilient Tapes and Treads 200 Series
Click to view available options
Color:
Transparent
Unit Size:
1 roll
Description
- For wet, barefoot-traffic areas
- Non-mineral, fine-textured, slip-resistant vinyl surface
- High performance, water-resistant, acrylate adhesive is ideal for wet conditions
- Applications: showers, tubs, surfboards, around hot tubs and pools
- Custom shapes and sizes are available
- Fine-textured, slip-resistant surface with a pressure-sensitive adhesive backing provides enhanced safety
- Custom shapes and sizes are available
Specifications
Specifications
| For Use With (Application) | Wet, Barefoot Traffic Areas |
| Length (Metric) | 18.288 m |
| Width (Metric) | 1219.2 mm |
| Color | Transparent |