Description
Securing joints by using our KECK™-products is a well-known and proven method. In order to meet high safety demands, e.g. when dealing with hazardous substances or pressure differences, DURAN™ Safety Joints offer very good protection against accidental loosening of the connection. Furthermore, DURAN™ Safety Joints offer a system to separate jammed cones and sockets. This minimizes the risk of breakage and related threats. The DURAN™ threaded sleeves also offer secure locking with DURAN™ standard cores.
- Threaded sleeve prevents unintentional loosening of the ground joint
- Firmly seated ground joints can be loosened more easily here. The danger of glass breakage and associated risks is thus minimized.
- Use in equipment for handling hazardous substances or working with pressure differences.g in the high vacuum range
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifications
Specifications
| Quantity | 10/Pk. |
| Item Description | Use in equipment for handling hazardous substances or working with pressure differences in the high vacuum range; When working under increased safety requirements; Installation in glass apparatus and devices |
| Joint Size | 18 mm |
| Thickness (Metric) Wall | 1.8 mm |