Learn More
DWK Life Sciences Safety Standard Ground Socket, with thread
Product Code. 15826921 Shop All DWK Life Sciences Products
Change view
Click to view available options
Joint Size:
18 mm
22 mm
28 mm
34 mm
50 mm
Unit Size:
Pack of 10
Product Code. Joint Size unitSize
15826921 50 mm Pack of 10
15806921 34 mm Pack of 10
15876911 18 mm Pack of 10
15886911 22 mm Pack of 10
15896911 28 mm Pack of 10
5 options
Product Code. 15826921
Brand: DWK Life Sciences 286594504
Only null left
Add to basket
Add to basket
DURAN™ safety ground joints for increased safety requirements.
Securing joints by using our KECK™-products is a well-known and proven method. In order to meet high safety demands, e.g. when dealing with hazardous substances or pressure differences, DURAN™ Safety Joints offer very good protection against accidental loosening of the connection. Furthermore, DURAN™ Safety Joints offer a system to separate jammed cones and sockets. This minimizes the risk of breakage and related threats. The DURAN™ threaded sleeves also offer secure locking with DURAN™ standard cores.
  • Threaded sleeve prevents unintentional loosening of the ground joint
  • Firmly seated ground joints can be loosened more easily here. The danger of glass breakage and associated risks is thus minimized.
  • Use in equipment for handling hazardous substances or working with pressure differences.g in the high vacuum range
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10/Pk.
Item Description Use in equipment for handling hazardous substances or working with pressure differences in the high vacuum range; When working under increased safety requirements; Installation in glass apparatus and devices
Joint Size 50 mm
Thickness (Metric) Wall 2.5 mm