Learn More
DWK Life Sciences Safety Standard Ground Socket, with thread
Product Code. 15806921 Shop All DWK Life Sciences Products
Click to view available options
Joint Size:
18 mm
22 mm
28 mm
34 mm
50 mm
Unit Size:
Pack of 10
Product Code. 15806921
Brand: DWK Life Sciences 286592906
Only null left
Add to basket
Add to basket
DURAN™ safety ground joints for increased safety requirements.
Securing joints by using our KECK™-products is a well-known and proven method. In order to meet high safety demands, e.g. when dealing with hazardous substances or pressure differences, DURAN™ Safety Joints offer very good protection against accidental loosening of the connection. Furthermore, DURAN™ Safety Joints offer a system to separate jammed cones and sockets. This minimizes the risk of breakage and related threats. The DURAN™ threaded sleeves also offer secure locking with DURAN™ standard cores.
  • Threaded sleeve prevents unintentional loosening of the ground joint
  • Firmly seated ground joints can be loosened more easily here. The danger of glass breakage and associated risks is thus minimized.
  • Use in equipment for handling hazardous substances or working with pressure differences.g in the high vacuum range
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10/Pk.
Item Description Use in equipment for handling hazardous substances or working with pressure differences in the high vacuum range; When working under increased safety requirements; Installation in glass apparatus and devices
Joint Size 34 mm
Thickness (Metric) Wall 2 mm