Learn More
Brady™ Safety Lockout
Product Code. 15653537
Change view
Click to view available options
Width (Metric) Shackle:
25.00 mm
38.00 mm
Unit Size:
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Shackle unitSize
15653537 25.00 mm Pack of 12
15663537 38.00 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15653537 Supplier Brady™ Supplier No. 805840
  • ONLINE EXCLUSIVE PRICE
    1285.00 NOK
    1193.53 NOK / Pack of 12
    Save 91.47 NOK
    7% Off
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
Machinery stays locked until every individual working on the repairs or inspection is finished.
  • Each person puts a lock on the hasp, but the hasp cannot be removed until all lockouts have been removed, thus preventing accidents
Specifications
Alternate Part Number 065375
Color Red
Material Vinyl-coated High Tensile Steel
Description Safety Lockout
Height (Metric) 129.29 mm
Quantity 12 Pk.
Width (Metric) 58.42 mm
Width (Metric) Shackle 25.00 mm