Learn More
3M™ Safety Glasses Carrying Case
Product Code. 15789677
Change view
Click to view available options
Style:
Rigid
Semi Rigid
Soft with Zipper
Unit Size:
Each
Pack of 20
Product Code. Style unitSize
15789677 Soft with Zipper Pack of 20
15799807 Rigid Each
15709817 Semi Rigid Pack of 20
3 options
Product Code. 15789677
Brand: 3M™ 7000032497
Only null left
Add to basket
Add to basket
Protect your safety eyewear from being scratched or damaged with the Safety Glasses Carrying Case. Ideal for protecting your safety glasses while they’re not in use.
  • Helps protect your 3M™ Safety Glasses and improves lifetime.
  • Choose from several styles:
    • Rigid safety glasses carrying case features a snap clasp and belt loop that allows you to easily carry your glasses.
    • Semi-rigid safety glasses carrying case with belt clip for attaching to belt loops.
    • Soft safety glasses carrying case features a zipper, belt loop, and snap clasp.
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Product Type Eyewear Case
For Use With (Equipment) Safety Glasses
Includes Zipper, belt loop, snap clasp
Model 12-0600-00
Style Soft with Zipper