Learn More
SCAT™ Safety Funnel 'ARNOLD' Replacement Lid
Shop All SCAT Europe Products
Click to view available options
Color:
Black
Blue, White
Material:
HDPE
HDPE-EC
Unit Size:
Each
Description
- Spare/replacement lid for use with the 'ARNOLD' V2.0 Safety Funnel
Specifications
Specifications
| Material | HDPE-EC |
| Height (Metric) | 5 mm |
| Color | Black |
| For Use With (Equipment) | ARNOLD Funnel |
| Includes | Replacement Lid |
| Product Type | Funnel Lid |