Description
- Made from tough clear acrylic for maximum visibility
- Prevents accidental touching of a hot block or contamination of samples from splashes
Specifications
Specifications
| For Use With (Equipment) | Fisherbrand™ 4 Block System |
| Type | Safety Cover |
| For Use With (Application) | Preventing Burns or Contamination |
| Material | Acrylic |
| Height (Metric) | 85 mm |