Learn More
Justrite™ Safety Cabinet Compact
Product Code. 11975416
Product Code. 11975416
Brand: Justrite™ 8912021
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
with rollers & easel not for phenol, nitric /sulphuric acid, steel, double wall, epoxy powder finish, dual vents, earth wire connections, leakproof sills, levelling feet blue 889mm x 591mm x 457mm (w x h x d) 45L