Learn More
Justrite™ Safety Cabinet 2 Door
Product Code. 11925436
Product Code. 11925436 Supplier Justrite™ Supplier No. 8960221
  • ONLINE EXCLUSIVE PRICE
    31652.07 NOK
    25855.26 NOK / Pack of 1
    Save 5796.81 NOK
    18% Off
 Restricted Item
Estimated Shipment: 14-05-2027
Self close, 2 adj shelves, not for phenol, nitric /sulphuric acid, steel, double wall dual vents, earth wire connections, leakproof sills, levelling feet blue 1,651mm x 864mm x 864mm (w x h x d) 227L