Description
Composed of flash spun high density polyethylene. Dupont™ Tyvek™ Classic Xpert Hooded Coverall is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
- Robust yet lightweight
- 3-piece hood for optimal fit to head and face when turning
- Elasticated face, wrists and ankles as well as glued-in waist elastic
- Ample crotch area for freedom of movement
- Large, easy-to-grasp zipper puller
- Very low inward leakage thanks to optimized design
Specifications
Specifications
| Size | 4X-Large |
| Color | White |
| Closure Type | Zipper |
| Seam Type | Stitched |
| Certifications/Compliance (Europe) | (EU) 2016/425, EN 14126, EN 1149-5 |
The user should make the risk assessment to determine the protection required.