Learn More
MEDICOM Safemask™ Fit+Pro FFP3 NR Valve Mask
Product Code. 31781311 Shop All Medicom Products
Change view
Click to view available options
Size:
One Size
Unit Size:
Pack of 144
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781311 One Size Pack of 144
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31781311 Supplier MEDICOM Supplier No. M63110V
Estimated Shipment: 05-10-2026
SafeMask Fit+Pro FFP3 valved mask protects against toxic aerosols and high concentrations, providing enhanced breathing comfort.
  • Folded 3-panel design adapts to various face shapes and sizes
  • Sealing foams on both the upper and lower panels ensure an ideal seal while providing unmatched comfort for the wearer
  • Easy to put on and adjust thanks to sliding elastic straps and a tightening ring
  • The exhalation valve effectively expels heat and humidity, reducing the risk of fogging on the wearer's glasses
  • Its individual packaging helps control the risk of contamination
Specifications
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type FFP3
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve Yes