Learn More
Brady™ SafeKey Padlocks - Nylon with 38 mm Plastic Shackle, Keyed Different
Product Code. p-7214025
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
White
Yellow
Quantity:
1 Ea.
6 Pk.
Unit Size:
Each
Pack of 6
Product Code. Color Quantity unitSize
17170281 Orange 1 Ea. Each
17180281 Black 1 Ea. Each
17190281 Yellow 1 Ea. Each
17180311 Purple 1 Ea. Each
17190311 Green 1 Ea. Each
17170351 Brown 1 Ea. Each
17140371 Red 1 Ea. Each
17190381 White 1 Ea. Each
17100391 Blue 1 Ea. Each
17100281 Blue 6 Pk. Pack of 6
17120311 Yellow 6 Pk. Pack of 6
17190341 White 6 Pk. Pack of 6
17100351 Brown 6 Pk. Pack of 6
17110351 Orange 6 Pk. Pack of 6
17120351 Red 6 Pk. Pack of 6
17190361 Green 6 Pk. Pack of 6
17110381 Purple 6 Pk. Pack of 6
17120381 Black 6 Pk. Pack of 6
18 options
Product Code. 17170281
Brand: Brady™ 150230
The ultimate locks for all of your safety and security needs
  • Compliments the Brady™ SafeKey Padlock range and have the same features
  • The new designed and innovative keying mechanism provides you much more unique key possibilities than a conventional standard safety padlock, without sacrificing a secure lockout
  • The unique lock mechanism is insulated and non-conductive to protect workers from shocks when key is inserted
  • The lock mechanism has 6 precision steel tumbler blades for a high number of key differences and tamper resistance
  • Locks are lightweight and resist impact, temperature, chemicals and corrosion
  • Their non-conductive, non-sparking body features a ribbed design for easier gripping and handling
  • The colour-matched key and lock body allows a fast identification in the application
  • 1 key supplied with each lock
  • Key retaining (Key won’t release until the padlock shackle is securely closed)
  • Custom keying, engraving, charting, Master Key and Grand Master Key are available
Specifications
Product Type Keyed Padlocks
Key Type Keyed Different
Alternate Part Number NYL-ORG-38PL-KD
Color Orange
Material Nylon
Diameter (English) Shackle 0.25 in.
Diameter (Metric) Shackle 6.4 mm
Height (English) Shackle 1.5 in.
Height (Metric) Shackle 38 mm
Material (Shackle) Plastic
No. of Pins 6
Quantity 1 Ea.
Width (English) 1.5 in.
Width (English) Shackle 1.3 in.
Width (Metric) 38 mm
Width (Metric) Shackle 33 mm
Show More Show Less
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.