Description
- Safe Sachet Handling
- The rotating sachet holder sits securely on the EasyPlace baseplate. It holds up to three standard sachets, preventing spills
- Straightforward Calibration
- The placement on the EasyPlace makes sensor insertion simple. A simple turn of the holder moves the next standard into the measurement position
Specifications
Specifications
| Type | Sachet Holder, EasyPlace Rotating |
| For Use With (Equipment) | EasyPlace |