Learn More
SAC3D1 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25857-1-AP-150UL
Additional Details : Weight : 0.00020kg
 View more versions of this product
Product Code. 17201386
  • 4593.76 NOK / 150µL
Estimated Shipment: 23-05-2024
Description
The SAC3D1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human SAC3D1. This antibody recognizes human antigen. The SAC3D1 antibody has been validated for the following applications: IHC, WB,ELISA analysis.
Specifications
Specifications
SAC3D1
Polyclonal
Unconjugated
150 μL
29901
Liquid
Western Blot, Immunohistochemistry, ELISA
450 μg/mL