missing translation for 'onlineSavingsMsg'
Learn More

Melford Sabouraud Dextrose Agar

Product Code. 30018767
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018767 1 kg 1kg
30018768 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018767 Supplier Melford Supplier No. S125001000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sabouraud Dextrose Agar is an agar medium recommended for the isolation, cultivation and identification of non-pathogenic and pathogenic fungi.

Sabouraud Dextrose Agar is an agar medium recommended for the isolation, cultivation and identification of non-pathogenic and pathogenic fungi. This medium does not contain antibiotics or inhibitors and can be used for the cultivation of those fungi which are susceptible to antibiotics.

Dissolve 65.0 grams in 1 liter distilled water. Sterilize by autoclaving for 15 minutes.

Ingredients:

Casein Digest Peptone: 10.0 g/L

Dextrose: 40.0 g/L

Agar: 15.0 g/L

Specifications

Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.