Learn More
SAA4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20398-1-AP-150UL
 View more versions of this product
Product Code. 16849934
  • 4836.10 NOK / 150µL
Description
SAA4 is a major acute phase reactant. It is an Apolipoprotein of the HDL complex.
Specifications
Specifications
SAA4
Polyclonal
Unconjugated
SAA4
C SAA, CSAA, SAA4, Serum amyloid A 4 protein, serum amyloid A4, constitutive
Rabbit
Antigen Affinity Chromatography
RUO
6291
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P35542
SAA4
SAA4 Fusion Protein Ag14081