Learn More
Brady™ S3700 Multicolor and Cut, Sign and Label Printer, with SFIDS
Color your way to an improved workplace with the S3700 Sign and Label Printer. With multicolor print and shape cutting capabilities, this printer creates impactful identification that fits your application and workspace needs.
40393.72 NOK
Specifications
Product Type | Printer |
---|---|
Color | Blue, Gray |
Display Type | Color LCD Touchscreen |
For Use With (Equipment) | Brady Labels |
Material | Plastic |
Product Code | Brand | Keyboard Type | Connector Types | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Keyboard Type | Connector Types | Description | Price | Quantity & Availability | |||
17862669
|
Brady™
321264 |
QWERTZ | Ethernet, Standalone Use, USB, Wi-Fi, Bluetooth™ 5 Low Energy (class II) | Printer with QWERTZ keyboard, Wifi and Bluetooth enabled and BWS SFIDS |
40393.72 NOK
1 set |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17882669
|
Brady™
321265 |
QWERTZ | Standalone Use, USB, Ethernet | Printer with QWERTZ keyboard and BWS SFIDS |
40393.72 NOK
1 set |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17892659
|
Brady™
321266 |
AZERTY | Ethernet, Standalone Use, USB, Wi-Fi, Bluetooth™ 5 Low Energy (class II) | Printer with AZERTY keyboard, Wifi and Bluetooth enabled and BWS SFIDS |
40393.72 NOK
1 set |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17822669
|
Brady™
321267 |
AZERTY | Standalone Use, USB, Ethernet | Printer with AZERTY Keyboard with BWS SFIDS |
40393.72 NOK
1 set |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17892669
|
Brady™
321268 |
QWERTY (EU) | Ethernet, Standalone Use, USB, Wi-Fi, Bluetooth™ 5 Low Energy (class II) | Printer with QWERTY EU keyboard, Wifi and Bluetooth enabled and BWS SFIDS |
40393.72 NOK
1 set |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17882679
|
Brady™
321269 |
QWERTY (EU) | Standalone Use, USB, Ethernet | Printer - QWERTY EU keyboard with BWS SFIDS |
40393.72 NOK
1 set |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17812679
|
Brady™
321270 |
QWERTY (UK) | Ethernet, Standalone Use, USB, Wi-Fi, Bluetooth™ 5 Low Energy (class II) | Printer with QWERTY UK keyboard, Wifi and Bluetooth enabled and BWS SFIDS |
40393.72 NOK
1 set |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17812669
|
Brady™
321271 |
QWERTY (UK) | Standalone Use, USB, Ethernet | Printer with QWERTY UK keyboard with BWS SFIDS |
40393.72 NOK
1 set |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17802679
|
Brady™
321272 |
CYRILLIC | Ethernet, Standalone Use, USB, Wi-Fi, Bluetooth™ 5 Low Energy (class II) | Printer with CYRILLIC keyboard, Wifi and Bluetooth enabled and BWS SFIDS |
40393.72 NOK
1 set |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17892679
|
Brady™
321273 |
CYRILLIC | Standalone Use, USB, Ethernet | Printer with CYRILLIC keyboard with BWS SFIDS |
40393.72 NOK
1 set |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Color your way to an improved workplace with the S3700 Sign and Label Printer. With multicolour print and shape cutting capabilities, this printer creates impactful identification that fits your application and workspace needs.
- Multicolor print capabilities make it ideal for creating impactful safety signs, lockout tags, warning labels and more in a single pass
- Built in XY-cutter allows for the creation of die-cut shapes and text that gives the flexibility to choose where to place new messaging
- The built-in keyboard and touchscreen interface make label creation a breeze
- Label creation is even faster with the quick setup and print speeds that come with print smart technology from Brady
- Prints on a variety of material options so you can create signs and labels for a variety of applications
- Includes Brady Workstation Safety and Facility ID Software Suite to easily create custom signs, labels and pipemarkers.
Specifications
Printer | |
Color LCD Touchscreen | |
Plastic | |
14.25 in. | |
361.95 mm | |
11 in. | |
Multicolor, Monocolor | |
S3700 Safety and Facility Printer, Brady Workstation Safety and Facility ID Software, Quick Start Guide, Power Cord, USB Cord, Mouse, Cleaning Kit (PCK-6), USB Flash Drive | |
512 MB | |
127.00 mm/sec. | |
36.9 lb. | |
cULus, CE, UL-AR and S Mark, UL NOM CoC, RCM Mark, EAC, KC, CCC, RoHS, BIS, BSMI, LOA, SABS | |
1 Ea. | |
AC Adaptor |
Blue, Gray | |
Brady Labels | |
Thermal Transfer | |
19 in. | |
482.60 mm | |
279.40 mm | |
155.19 x 86.87 mm | |
English (US), French, Portuguese, Spanish, Bahasa (Indonesian), Bahasa Malay, Czech, Danish, German, Estonian, Croatian, Italian, Hungarian, Dutch, Norwegian (Bokmål), Polish, Romanian, Slovak, Slovenian, Finnish, Swedish, Vietnamese, Turkish, Bulgarian, Russian, Korean, Chinese (Traditional and Simplified), Thai, Japanese | |
S3700 | |
300 dpi | |
16.74 kg | |
Asset tracking, Cold temperature labelling, Component and equipment labelling, Electrical labelling, Facility identification, General identification, Hazard communication identification, Lean and 5S labelling, Pipe and valve marking, Safety identification, Security identification, Temperature in.dicating labelling, Barcode labelling, Exit and egress marking, Floor marking, Food and Beverage Process labelling, Healthcare and Hospital labelling, Inventory and in.spection labelling, Temporary labelling, Warehouse marking | |
1 year | |
110/240 V |