Learn More
Tygon S3™ E-3603 Vacuum Tubing
Product Code. 30053914 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1 in.
1/2 in.
1/4 in.
3/16 in.
3/4 in.
3/8 in.
5/8 in.
Diameter (English) Outer:
1-1/2 in.
1-1/8 in.
1-3/8 in.
2 in.
5/8 in.
7/8 in.
9/16 in.
Unit Size:
15m
Product Code. Diameter (English) Inner Diameter (English) Outer unitSize
30053914 5/8 in. 1-3/8 in. 15m
30053203 3/4 in. 1-1/2 in. 15m
30053637 1/2 in. 1-1/8 in. 15m
30053573 1/4 in. 5/8 in. 15m
30054222 3/8 in. 7/8 in. 15m
30053333 1 in. 2 in. 15m
30053233 3/16 in. 9/16 in. 15m
7 options
Product Code. 30053914
Brand: Tygon 5011369
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Formulated to resist flex-fatigue and abrasion
Bio-based formulation reduces environmental impact. Lot-to-lot consistency for reproducible results. Easily slips over fittings and grips securely. Contains no BPA or phthalates. Meets all USP Class VI and FDA food contact criteria. . Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Specifications
Certifications/Compliance USP;FDA
Diameter (English) Inner 5/8 in.
Material Tygon S3 E-3603
Diameter (English) Outer 1-3/8 in.