missing translation for 'onlineSavingsMsg'
Learn More

S3-12 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55404-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16846865

  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

S3-12
Polyclonal
Unconjugated
PLIN4
Adipocyte protein S3 12, KIAA1881, Perilipin 4, PLIN4, S3 12
Rabbit
Antigen Affinity Chromatography
RUO
729359
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.45 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96Q06
PLIN4
Peptide
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers