missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S3-12 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55404-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| S3-12 | |
| Polyclonal | |
| Unconjugated | |
| PLIN4 | |
| Adipocyte protein S3 12, KIAA1881, Perilipin 4, PLIN4, S3 12 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 729359 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96Q06 | |
| PLIN4 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction