missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S1P2/EDG-5/S1PR2 Antibody [Janelia Fluor« 525], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
S1P2/EDG-5/S1PR2 Polyclonal antibody specifically detects S1P2/EDG-5/S1PR2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | S1P2/EDG-5/S1PR2 |
| Applications | ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 525 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | AGR16, EDG5, EDG-5, endothelial differentiation G-protein coupled receptor 5, endothelial differentiation, sphingolipid G-protein-coupled receptor, 5, Gpcr13, H218, LPB2, S1P receptor 2, S1P receptor Edg-5, S1P receptor EDG5, S1P2, sphingosine 1-phosphate receptor 2, sphingosine 1-phosphate receptor Edg-5, sphingosine-1-phosphate receptor 2 |
| Host Species | Rabbit |
| Immunogen | S1P2/EDG-5/S1PR2 antibody was raised against an 16 amino acid peptide near the carboxy terminus of human S1P2/EDG-5/S1PR2. The immunogen is located within amino acids 270 - 320 of S1P2/EDG-5/S1PR2. |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?