Description
Reduce separation time, processing samples 17% faster than previous similar rotors at an RCF of 691,000 x g using the Thermo Scientific™ S110-AT Fixed Angle Rotor.
Compatible with:
Thermo Scientific™ Sorvall™ MTX 150 Series Micro-Ultracentrifuge, MX Series Floor Model Micro-Ultracentrifuge
Specifications
Specifications
| Description | S110-AT Fixed Angle Rotor |
| Material | Titanium |
| Type | Fixed Angle Rotor |
| Capacity | 8 x 5 mL |
| Max. RCF | 690,652 x g |
| Max. Speed | 110,000 rpm |
| Radius (Metric) | 51.1 mm |
| Radius (Metric) Min. | 24.5 mm |
| K Factor | 15.4 |
| Max. Tube Length | 63 mm |
| Show More |