missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100G Antibody (S100G/7461), Novus Biologicals™
Mouse Monoclonal Antibody
2100.00 NOK - 4170.00 NOK
Specifications
| Antigen | S100G |
|---|---|
| Clone | S100G/7461 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619414
|
Novus Biologicals
NBP3-23850-100ug |
100 μg |
4170.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663884
|
Novus Biologicals
NBP3-23850-20ug |
20 μg |
2100.00 NOK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S100G Monoclonal antibody specifically detects S100G in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| S100G | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| CABP, CABP1, CaBP9K, CABP9KMGC138379, CALB3calbindin D9K, calbindin 3, (vitamin D-dependent calcium-binding protein), calbindin-D9K, protein S100-G, S100 calcium binding protein G, S100 calcium-binding protein G, S100D, Vitamin D-dependent calcium-binding protein, intestinal | |
| Recombinant fragment (around aa1-79) of human S100G protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| S100G/7461 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 795 | |
| IgG2 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title