missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A7/Psoriasin Antibody (030), DyLight 488 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89812G
This item is not returnable.
View return policy
Description
S100A7/Psoriasin Monoclonal antibody specifically detects S100A7/Psoriasin in Human samples. It is validated for ELISA
Specifications
| S100A7/Psoriasin | |
| Monoclonal | |
| DyLight 488 | |
| HID 5, HID5, HID-5, protein S100-A7, PSOR1, Psoriasin, psoriasin 1, S100 calcium binding protein A7, S100 calcium binding protein A7 (psoriasin 1), S100 calcium-binding protein A7, S100 calcium-binding protein A7 (psoriasin 1), S100A7c | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human S100A7/Psoriasin (Met 1-Gln 101; NP_002954.2). | |
| 0.1 mL | |
| Cancer | |
| 6278 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 030 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction