Learn More
S0859, MedChemExpress
Product Code. 16469167
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16469167
Brand: Medchem Express HY15529
MedChemExpress S0859 is a selective, high-affinity generic Na+/HCO3- transporter (NBC) inhibitor. S0859 reversibly inhibits NBC-mediated intracellular pH (pHi) recovery (Ki=1.7 μM, full inhibition at approximately 30 μM).
S0859 is a selective, high-affinity generic Na+/HCO3- transporter (NBC) inhibitor. S0859 reversibly inhibits NBC-mediated intracellular pH (pHi) recovery (Ki=1.7 μM, full inhibition at approximately 30 μM).
Specifications
Chemical Name or Material S0859
CAS 1019331-10-2
Color White
Purity Grade Notes Research
Molecular Formula C29H24ClN3O3S
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (188.67 mM)
SMILES O=C(N(CC1=CC=C(C2=CC=CC=C2S(=O)(NC#N)=O)C=C1)CC3=CC=C(C)C=C3)C4=CC=CC=C4Cl
Molecular Weight (g/mol) 530.04
Formula Weight 530.04
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.