Learn More
(-)-S-Trityl-D-cysteine, 98%
Product Code. p-7054646
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15469458 1 g 1g
15479458 5 g 5g
2 options
Product Code. 15469458
Brand: Thermo Scientific Alfa Aesar H63022.03
Request Bulk or Custom Format
CAS: 25840-82-8 | C22H21NO2S | 363.48 g/mol
(-)-S-Trityl-D-cysteine is used in medicine as intermediate. It inhibits the human mitotic kinesin Eg5.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(-)-S-Trityl-D-cysteine is used in medicine as intermediate. It inhibits the human mitotic kinesin Eg5.
Solubility
Sparingly soluble in water 0.022 g/L 25°C.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25840-82-8
Molecular Formula C22H21NO2S
Molecular Weight (g/mol) 363.48
MDL Number MFCD00236948
InChI Key DLMYFMLKORXJPO-UHFFFAOYNA-N
Synonym h-d-cys trt-oh, s-trityl-d-cysteine, h-cys trt-oh, s-2-amino-3-tritylthio propanoic acid, 2s-2-amino-3-triphenylmethyl sulfanyl propanoic acid, d-cysteine, s-triphenylmethyl, ambotzhaa6120, h-cys trt-2-chlorotrityl resin, --s-trityl-d-cysteine, 2s-2-amino-3-tritylsulfanylpropanoic acid
PubChem CID 7271796
IUPAC Name 2-amino-3-[(triphenylmethyl)sulfanyl]propanoic acid
SMILES NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
Specifications
Solubility Information Sparingly soluble in water 0.022g/L 25°C.
Quantity 1 g
Formula Weight 363.47
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (-)-S-Trityl-D-cysteine