Learn More
S protein RBD anti-SARS-CoV-2, Clone: AM180, ACROBiosystems
Description
This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. Cross-reactivity with RBD of other coronaviruses has not been tested.
Specifications
Specifications
| Antigen | S protein RBD |
| Applications | ELISA |
| Clone | AM180 |
| Conjugate | Unconjugated |
| Formulation | Lyophilized from 0.22μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization. |
| Host Species | Human (HEK293) |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Test Specificity | This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. Cross-reactivity with RBD of other coronaviruses has not been tested. |
| Show More |